siRNA Details
Gene Symbol: C6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176074.3 | NP_788263.1 | 1, 2 | 144 | ||
XM_008760747.1 | XP_008758969.1 | 2, 3 | 333 | |||
XM_008760749.1 | XP_008758971.1 | 2, 3 | 318 | |||
XM_008760750.1 | XP_008758972.1 | 2, 3 | 332 | |||
GenBank mRNA | AY230250.1 | - | 1, 2 | 146 |
