siRNA Details
Gene Symbol: Zfhx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098803.1 | NP_001092273.1 | 9 | 4927 | ||
XM_008770677.1 | XP_008768899.1 | 8 | 6702 | |||
XM_008770678.1 | XP_008768900.1 | 7 | 6627 | |||
GenBank mRNA | AY237644.1 | - | 1 | 237 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
