siRNA Details
Gene Symbol: Oxgr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207588.1 | NP_997471.1 | 1 | 208 | ||
XM_006252471.2 | XP_006252533.1 | 2 | 458 | |||
GenBank mRNA | AY191367.1 | - | 1 | 208 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Oxgr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207588.1 | NP_997471.1 | 1 | 208 | ||
XM_006252471.2 | XP_006252533.1 | 2 | 458 | |||
GenBank mRNA | AY191367.1 | - | 1 | 208 |