siRNA Details
Gene Symbol: Nmt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207590.1 | NP_997473.1 | 11 | 1134 | ||
XM_008771854.1 | XP_008770076.1 | 11 | 1153 | |||
XM_008771855.1 | XP_008770077.1 | 9 | 1156 | |||
XM_008771856.1 | XP_008770078.1 | 10 | 1102 | |||
GenBank mRNA | AJ629217.1 | - | 11 | 1134 |
