siRNA Details
Gene Symbol: Ankra2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207595.1 | NP_997478.1 | 9 | 1459 | ||
GenBank mRNA | AY566568.1 | - | 9 | 1459 |
Gene Symbol: LOC100910438 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008760690.1 | XP_008758912.1 | 9 | 1710 | ||
XM_008760691.1 | XP_008758913.1 | 8 | 1264 |
