siRNA Details
Gene Symbol: Ns5atp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207607.1 | NP_997490.1 | 1 | 615 | ||
GenBank mRNA | AY533136.1 | - | 1 | 615 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
