siRNA Details
Gene Symbol: MIEF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304564.1 | NP_001291493.1 | 7 | 4221 | ||
NM_019008.5 | NP_061881.2 | 6 | 5276 | |||
NR_130789.1 | 6 | 5190 | ||||
XM_011530236.1 | XP_011528538.1 | 8 | 5344 | |||
XM_011530237.1 | XP_011528539.1 | 7 | 5464 | |||
XM_011530238.1 | XP_011528540.1 | 7 | 5548 | |||
XM_017028838.1 | XP_016884327.1 | 7 | 5442 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
