siRNA Details
Gene Symbol: GRK6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004106.2 | NP_001004106.1 | 16 | 2901 | ||
NM_002082.3 | NP_002073.2 | 16 | 2903 | |||
GenBank mRNA | L16862.1 | - | 16 | 2796 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
