siRNA Details
Gene Symbol: GHRHR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000823.3 | NP_000814.2 | 12, 13 | 1196 | ||
XM_011515263.1 | XP_011513565.1 | 9, 10 | 1214 | |||
GenBank mRNA | AF282259.1 | - | 9, 10 | 1214 | ||
AF282260.1 | - | 8, 9 | 1060 | |||
AF282261.1 | - | 9, 10 | 884 | |||
AF282262.1 | - | 6, 7 | 829 | |||
AY557192.1 | - | 12, 13 | 1148 | |||
L01406.1 | - | 12, 13 | 1199 | |||
L09237.1 | - | 12, 13 | 1196 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
