siRNA Details
Gene Symbol: AGGF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018046.4 | NP_060516.2 | 1 | 467 | ||
GenBank mRNA | AY500994.1 | - | 1 | 386 | ||
BC002828.2 | - | 1 | 407 | |||
BC032844.1 | - | 1 | 453 | 5266978 | ||
BI561566.1 | - | 1 | 471 | |||
CF994178.1 | - | 472 | ||||
U84971.1 | - | 1 | 396 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
