siRNA Details
Gene Symbol: OR8B8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012378.1 | NP_036510.1 | 1 | 294 | ||
GenBank mRNA | AF238488.1 | - | 1 | 294 | ||
BC130393.1 | - | 1 | 367 | 40146424 | ||
BC130419.1 | - | 1 | 367 | 40146450 |
Gene Symbol: OR8B12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005195.1 | NP_001005195.1 | 1 | 291 | ||
GenBank mRNA | BC136863.1 | - | 1 | 316 | 9020854 | |
BC136864.1 | - | 1 | 316 | 9020855 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
