siRNA Details
Gene Symbol: OR7A17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030901.1 | NP_112163.1 | 1 | 668 | ||
GenBank mRNA | BC101587.1 | - | 1 | 765 | 8069093 | |
BC117357.1 | - | 1 | 765 | 40125908 | ||
X64993.1 | - | 1 | 294 |
Gene Symbol: OR7A10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005190.1 | NP_001005190.1 | 1 | 668 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
