siRNA Details
Gene Symbol: TIAM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012454.3 | NP_036586.2 | 8 | 2465 | ||
GenBank mRNA | AF120323.1 | - | 3 | 363 | ||
AK123859.1 | - | 9 | 2560 | |||
AK125459.1 | - | 5 | 924 | |||
AK302860.1 | - | 3 | 304 | |||
AL122086.1 | - | 3 | 294 | |||
BC137275.1 | - | 7 | 2338 | 9021272 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
