siRNA Details
Gene Symbol: DUSP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001947.3 | NP_001938.2 | 2 | 957 | ||
GenBank mRNA | AF508727.1 | - | 2 | 725 | ||
BC019107.2 | - | 2 | 589 | |||
BC104880.1 | - | 2 | 785 | 8143883 | ||
BC104882.1 | - | 2 | 785 | 8143885 | ||
X93921.1 | - | 2 | 587 |
