siRNA Details
Gene Symbol: RXFP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181885.2 | NP_871001.1 | 1 | 103 | ||
GenBank mRNA | AY170824.1 | - | 1 | 82 | ||
AY288415.1 | - | 1 | 82 | |||
AY394502.1 | - | 1 | 82 | |||
BC101507.1 | - | 1 | 102 | 8069013 | ||
BC101509.1 | - | 1 | 102 | 8069015 | ||
DN693323.1 | - | 1 | 103 | |||
EU432129.1 | - | 1 | 82 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
