siRNA Details
Gene Symbol: MAP3K9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284230.1 | NP_001271159.1 | 10, 11 | 2020 | ||
XM_005267683.4 | XP_005267740.1 | 9, 10 | 2046 | |||
GenBank mRNA | AY327900.1 | - | 10, 11 | 2020 | ||
BC111407.1 | - | 10, 11 | 2019 | |||
BC133706.1 | - | 10, 11 | 2020 | 40036384 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
