siRNA Details
Gene Symbol: AMY1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008221.1 | NP_001008222.1 | 5 | 926 | ||
NM_004038.3 | NP_004029.2 | 5 | 1007 | |||
GenBank mRNA | AK292341.1 | - | 5 | 925 | ||
AK313275.1 | - | 5 | 927 | |||
BC063129.1 | - | 5 | 936 | 30327425 | ||
BC069347.1 | - | 4 | 712 | 7262297 | ||
BC069463.1 | - | 4 | 712 | 7262309 | ||
BC070302.1 | - | 5 | 932 | |||
BC092444.1 | - | 5 | 903 | 30349929 | ||
BC132985.1 | - | 5 | 882 | 40147007 | ||
BC132987.1 | - | 5 | 882 | 40147009 | ||
BC132995.1 | - | 5 | 881 | 40147017 | ||
BC132997.1 | - | 5 | 882 | 40147019 | ||
BC144452.1 | - | 5 | 882 | 9052978 |
Gene Symbol: AMY1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008218.1 | NP_001008219.1 | 5 | 984 | ||
XM_011541262.1 | XP_011539564.1 | 5 | 927 | |||
GenBank mRNA | CB957894.1 | - | 3 | 525 | ||
CB958435.1 | - | 1 | 101 |
Gene Symbol: AMY1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008219.1 | NP_001008220.1 | 5 | 1006 | ||
XM_011541263.1 | XP_011539565.1 | 5 | 927 | |||
XM_017001058.1 | XP_016856547.1 | 5 | 844 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
