siRNA Details
Gene Symbol: OR4D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012374.1 | NP_036506.1 | 1 | 132 | ||
GenBank mRNA | BC136589.1 | - | 1 | 153 | 9020579 | |
BC136595.1 | - | 1 | 153 | 9020585 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
