siRNA Details
Gene Symbol: AMY2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020978.4 | NP_066188.1 | 3 | 750 | ||
GenBank mRNA | AK095605.1 | - | 2 | 1420 | ||
AK127047.1 | - | 2 | 2189 | |||
BC011179.1 | - | 3 | 442 | 4150646 | ||
BC020861.1 | - | 2 | 214 | 4551745 | ||
BX456665.2 | - | 3 | 694 | |||
M24895.1 | - | 1 | 124 |
Gene Symbol: AMY2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000699.3 | NP_000690.1 | 1 | 187 | ||
GenBank mRNA | AK223463.1 | - | 1 | 102 | ||
BC007060.1 | - | 1 | 142 | 3949799 | ||
BC146997.1 | - | 1 | 187 | 9056925 | ||
M28443.1 | - | 1 | 91 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
