siRNA Details
Gene Symbol: AMY1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008221.1 | NP_001008222.1 | 2, 3 | 390 | ||
NM_004038.3 | NP_004029.2 | 2, 3 | 471 | |||
GenBank mRNA | AK292341.1 | - | 2, 3 | 389 | ||
AK313275.1 | - | 2, 3 | 391 | |||
BC063129.1 | - | 2, 3 | 400 | 30327425 | ||
BC069347.1 | - | 1, 2 | 176 | 7262297 | ||
BC069463.1 | - | 1, 2 | 176 | 7262309 | ||
BC070302.1 | - | 2, 3 | 396 | |||
BC092444.1 | - | 2, 3 | 367 | 30349929 | ||
BC132985.1 | - | 2, 3 | 346 | 40147007 | ||
BC132987.1 | - | 2, 3 | 346 | 40147009 | ||
BC132995.1 | - | 2, 3 | 345 | 40147017 | ||
BC132997.1 | - | 2, 3 | 346 | 40147019 | ||
BC144452.1 | - | 2, 3 | 346 | 9052978 | ||
BX487266.1 | - | 2, 3 | 391 | |||
CB956272.1 | - | 2, 3 | 474 |
Gene Symbol: AMY1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008218.1 | NP_001008219.1 | 2, 3 | 448 | ||
XM_011541262.1 | XP_011539564.1 | 2, 3 | 391 | |||
GenBank mRNA | CB958705.1 | - | 2, 3 | 451 |
Gene Symbol: AMY1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008219.1 | NP_001008220.1 | 2, 3 | 470 | ||
XM_011541263.1 | XP_011539565.1 | 2, 3 | 391 | |||
XM_017001058.1 | XP_016856547.1 | 2, 3 | 308 | |||
GenBank mRNA | CB955582.1 | - | 2, 3 | 258 | ||
CB985420.1 | - | 2, 3 | 365 | |||
CB987307.1 | - | 2, 3 | 473 | |||
CD690933.1 | - | 3, 4 | 401 |
Gene Symbol: AMY2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000699.3 | NP_000690.1 | 1, 2 | 285 | ||
GenBank mRNA | AK223463.1 | - | 1, 2 | 200 | ||
BC007060.1 | - | 1, 2 | 240 | 3949799 | ||
BC146997.1 | - | 1, 2 | 285 | 9056925 | ||
M28443.1 | - | 1, 2 | 189 |
Gene Symbol: AMY2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020978.4 | NP_066188.1 | 3, 4 | 848 | ||
GenBank mRNA | AK095605.1 | - | 2, 3 | 1518 | ||
AK127047.1 | - | 2, 3 | 2287 | |||
BC011179.1 | - | 3, 4 | 540 | 4150646 | ||
BC020861.1 | - | 2, 3 | 312 | 4551745 | ||
BX456665.2 | - | 3, 4 | 792 | |||
M24895.1 | - | 1, 2 | 222 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
