siRNA Details
Gene Symbol: AKAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004065.4 | NP_001004065.2 | 2 | 366 | ||
NM_001136562.2 | NP_001130034.1 | 2 | 204 | |||
NM_001198656.1 | NP_001185585.1 | 2 | 366 | |||
GenBank mRNA | AK057098.1 | - | 2 | 268 | ||
AK300427.1 | - | 2 | 179 | |||
AK304311.1 | - | 2 | 248 | |||
AK308388.1 | - | 2 | 328 | |||
BC146863.1 | - | 2 | 366 | 9056791 | ||
BC171800.1 | - | 2 | 366 | 9054454 | ||
BE674805.1 | - | 2 | 165 | |||
DC422367.1 | - | 2 | 204 |
Gene Symbol: PALM2-AKAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007203.4 | NP_009134.1 | 8 | 882 | ||
NM_147150.2 | NP_671492.1 | 8 | 882 | |||
GenBank mRNA | AB023137.1 | - | 8 | 882 | ||
AJ303079.1 | - | 8 | 870 | |||
BC140818.1 | - | 8 | 723 | 9021686 | ||
BX649126.1 | - | 6 | 668 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
