siRNA Details
Gene Symbol: MMP23B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006983.1 | NP_008914.1 | 7 | 1015 | ||
XM_017002615.1 | XP_016858104.1 | 6 | 1923 | |||
XM_017002616.1 | XP_016858105.1 | 6 | 1923 | |||
XM_017002617.1 | XP_016858106.1 | 6 | 1923 | |||
GenBank mRNA | AB010961.1 | - | 7 | 1004 | ||
AB010962.1 | - | 6 | 1096 | |||
AF056200.1 | - | 7 | 992 | |||
AJ005256.1 | - | 7 | 1015 | |||
AK297700.1 | - | 5 | 1487 | |||
BC025719.1 | - | 7 | 1006 | 5228894 |
Gene Symbol: MMP23A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002946.1 | 5 | 684 | |||
GenBank mRNA | AF057061.1 | - | 4 | 658 | ||
AF057062.1 | - | 3 | 610 | |||
AF086353.1 | - | 4 | 430 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
