siRNA Details
Gene Symbol: CHKB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005198.4 | NP_005189.2 | 3 | 585 | ||
GenBank mRNA | AB029886.1 | - | 3 | 435 | ||
AK097775.1 | - | 3 | 753 | |||
AK314324.1 | - | 3 | 419 | |||
AL096780.1 | - | 3 | 510 | |||
AL096781.1 | - | 3 | 367 | |||
BC024286.1 | - | 3 | 364 | |||
BC082263.1 | - | 3 | 422 | 6384695 | ||
BC101488.1 | - | 3 | 430 | 8068994 | ||
BC113521.1 | - | 3 | 430 | 8322573 | ||
CR456419.1 | - | 3 | 496 |
Gene Symbol: CHKB-CPT1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027928.2 | 3 | 585 | |||
GenBank mRNA | AB051457.1 | - | 1 | 1137 | ||
AK301477.1 | - | 1 | 41 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
