siRNA Details
Gene Symbol: FFAR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005304.3 | NP_005295.1 | 2 | 1060 | ||
XM_011526858.2 | XP_011525160.1 | 2 | 1416 | |||
GenBank mRNA | AK290710.1 | - | 2 | 1060 | ||
BC035657.1 | - | 2 | 1046 | 5588752 | ||
BC113695.1 | - | 1 | 1055 | 8322747 | ||
BC148269.1 | - | 1 | 1007 | 8860212 | ||
EU432115.1 | - | 1 | 985 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
