siRNA Details
Gene Symbol: CELA3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007352.3 | NP_031378.1 | 6 | 754 | ||
GenBank mRNA | AK092382.1 | - | 8 | 549 | ||
AK315798.1 | - | 6 | 623 | |||
BC005216.1 | - | 6 | 628 | 3949903 | ||
BI760781.1 | - | 6 | 753 | |||
BM129235.1 | - | 5 | 534 | |||
M16630.1 | - | 6 | 599 | |||
M18692.1 | - | 6 | 626 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
