siRNA Details
Gene Symbol: CELA3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007352.3 | NP_031378.1 | 7 | 864 | ||
GenBank mRNA | AK092382.1 | - | 9 | 659 | ||
AK315798.1 | - | 7 | 733 | |||
BC005216.1 | - | 7 | 738 | 3949903 | ||
M16630.1 | - | 7 | 709 | |||
M18692.1 | - | 7 | 736 |
Gene Symbol: CELA3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005747.4 | NP_005738.4 | 7 | 737 | ||
GenBank mRNA | BI833796.1 | - | 2 | 187 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
