siRNA Details
Gene Symbol: KIR2DS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291695.1 | NP_001278624.1 | 3 | 244 | ||
NM_001291701.1 | NP_001278630.1 | 3 | 244 | |||
NM_012312.4 | NP_036444.1 | 3 | 244 | |||
GenBank mRNA | AF135565.1 | - | 1 | 146 | ||
AF285438.1 | - | 2 | 169 | |||
AJ002102.1 | - | 3 | 209 | |||
AJ002103.1 | - | 3 | 209 | |||
AJ002104.1 | - | 3 | 209 | |||
AJ002105.1 | - | 3 | 209 | |||
AM409094.1 | - | 3 | 210 | |||
AY366243.1 | - | 3 | 219 | |||
AY366244.1 | - | 3 | 208 | |||
BC108917.1 | - | 3 | 211 | 40017299 | ||
BC119631.1 | - | 3 | 228 | 40115568 | ||
BC119632.1 | - | 3 | 228 | 40115570 | ||
BC125146.1 | - | 2 | 193 | |||
BC126856.1 | - | 3 | 211 | 40017301 | ||
FR750994.1 | - | 3 | 244 | |||
L41347.1 | - | 3 | 228 | |||
L76667.1 | - | 3 | 220 | |||
U24079.1 | - | 3 | 207 | |||
X89893.1 | - | 3 | 209 |
Gene Symbol: KIR2DL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014219.2 | NP_055034.2 | 3 | 238 | ||
GenBank mRNA | AF285434.1 | - | 2 | 169 | ||
AY366242.1 | - | 3 | 238 | |||
KP260924.1 | - | 3 | 205 | |||
L76669.1 | - | 3 | 220 | |||
U24075.1 | - | 3 | 223 |
Gene Symbol: KIR2DL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015868.2 | NP_056952.2 | 3 | 246 | ||
GenBank mRNA | AF135566.1 | - | 1 | 146 | ||
AF135567.1 | - | 1 | 146 | |||
AF285435.1 | - | 2 | 169 | |||
AY789057.1 | - | 3 | 209 | |||
BC032422.1 | - | 3 | 222 | 5220339 | ||
BC050730.1 | - | 3 | 240 | 5219336 | ||
BC110613.1 | - | 3 | 223 | 40031619 | ||
FN395076.1 | - | 3 | 209 | |||
FN552550.1 | - | 3 | 191 | |||
L41268.1 | - | 3 | 246 | |||
L76662.1 | - | 3 | 220 | |||
L76663.1 | - | 3 | 220 | |||
U24074.1 | - | 3 | 230 | |||
U50546.1 | - | 3 | 220 | |||
U96190.1 | - | 1 | 146 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
