siRNA Details
Gene Symbol: KIR2DS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012313.1 | NP_036445.1 | 3 | 143 | ||
GenBank mRNA | AF022047.1 | - | 3 | 136 | ||
AM851002.1 | - | 3 | 168 | |||
AY789063.2 | - | 2 | 116 | |||
L76670.1 | - | 3 | 143 | |||
X97231.1 | - | 3 | 191 |
Gene Symbol: 1060P11.3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AJ630586.1 | - | 4 | 418 |
Gene Symbol: KIR2DL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014218.2 | NP_055033.2 | 3 | 170 | ||
XM_011526939.2 | XP_011525241.1 | 3 | 775 | |||
XM_017026782.1 | XP_016882271.1 | 3 | 148 | |||
XM_017026783.1 | XP_016882272.1 | 3 | 775 | |||
GenBank mRNA | AF022045.1 | - | 3 | 139 | ||
AF135560.1 | - | 1 | 69 | |||
AF285431.1 | - | 3 | 165 | |||
AF285432.1 | - | 2 | 92 | |||
AY366237.1 | - | 3 | 168 | |||
AY366238.1 | - | 3 | 114 | |||
AY366239.1 | - | 3 | 162 | |||
AY366240.1 | - | 3 | 170 | |||
AY366241.1 | - | 3 | 169 | |||
AY388471.1 | - | 3 | 162 | |||
AY789055.1 | - | 3 | 165 | |||
AY789056.1 | - | 3 | 132 | |||
BC069344.1 | - | 3 | 165 | 7262296 | ||
KJ699235.1 | - | 3 | 405 | |||
L41267.1 | - | 3 | 164 | |||
U24076.1 | - | 3 | 165 | |||
U24078.1 | - | 3 | 172 | |||
U96189.1 | - | 1 | 69 |
Gene Symbol: KIR2DL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014219.2 | NP_055034.2 | 3 | 161 | ||
GenBank mRNA | AF285434.1 | - | 2 | 92 | ||
AY366242.1 | - | 3 | 161 | |||
KP260924.1 | - | 3 | 128 | |||
L76669.1 | - | 3 | 143 | |||
U24075.1 | - | 3 | 146 |
Gene Symbol: KIR2DL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015868.2 | NP_056952.2 | 3 | 169 | ||
GenBank mRNA | AF022048.1 | - | 3 | 132 | ||
AF135556.1 | - | 1 | 69 | |||
AF135566.1 | - | 1 | 69 | |||
AF135567.1 | - | 1 | 69 | |||
AF285435.1 | - | 2 | 92 | |||
AY789057.1 | - | 3 | 132 | |||
BC032422.1 | - | 3 | 145 | 5220339 | ||
BC050730.1 | - | 3 | 163 | 5219336 | ||
BC110613.1 | - | 3 | 146 | 40031619 | ||
FN395076.1 | - | 3 | 132 | |||
FN552550.1 | - | 3 | 114 | |||
L41268.1 | - | 3 | 169 | |||
L76662.1 | - | 3 | 143 | |||
L76663.1 | - | 3 | 143 | |||
U24074.1 | - | 3 | 153 | |||
U50546.1 | - | 3 | 143 | |||
U73395.1 | - | 3 | 153 | |||
U96190.1 | - | 1 | 69 |
Gene Symbol: KIR2DS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014512.1 | NP_055327.1 | 3 | 145 | ||
GenBank mRNA | AF022046.1 | - | 3 | 145 | ||
AF285437.1 | - | 2 | 92 | |||
AM999888.1 | - | 3 | 140 | |||
X89892.1 | - | 3 | 152 | |||
X98858.1 | - | 3 | 132 |
Gene Symbol: KIR2DS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291695.1 | NP_001278624.1 | 3 | 167 | ||
NM_001291701.1 | NP_001278630.1 | 3 | 167 | |||
NM_012312.4 | NP_036444.1 | 3 | 167 | |||
GenBank mRNA | AF135565.1 | - | 1 | 69 | ||
AF285438.1 | - | 2 | 92 | |||
AJ002102.1 | - | 3 | 132 | |||
AJ002103.1 | - | 3 | 132 | |||
AJ002104.1 | - | 3 | 132 | |||
AJ002105.1 | - | 3 | 132 | |||
AM409094.1 | - | 3 | 133 | |||
AY366243.1 | - | 3 | 142 | |||
AY366244.1 | - | 3 | 131 | |||
BC108917.1 | - | 3 | 134 | 40017299 | ||
BC119631.1 | - | 3 | 151 | 40115568 | ||
BC119632.1 | - | 3 | 151 | 40115570 | ||
BC125146.1 | - | 2 | 116 | |||
BC126856.1 | - | 3 | 134 | 40017301 | ||
FR750994.1 | - | 3 | 167 | |||
L41347.1 | - | 3 | 151 | |||
L76667.1 | - | 3 | 143 | |||
U24079.1 | - | 3 | 130 | |||
X89893.1 | - | 3 | 132 |
Gene Symbol: KIR2DS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281971.1 | NP_001268900.1 | 3 | 221 | ||
NM_001281972.1 | NP_001268901.1 | 3 | 221 | |||
NM_012314.5 | NP_036446.3 | 3 | 221 | |||
GenBank mRNA | AF002255.1 | - | 3 | 178 | ||
AF135563.1 | - | 1 | 69 | |||
AF135564.1 | - | 1 | 69 | |||
AJ417554.1 | - | 2 | 112 | |||
AJ417555.1 | - | 2 | 112 | |||
AJ620615.2 | - | 2 | 105 | |||
AJ620616.2 | - | 2 | 105 | |||
AY102623.1 | - | 3 | 132 | |||
AY366245.1 | - | 3 | 166 | |||
AY366246.1 | - | 3 | 139 | |||
AY388472.1 | - | 3 | 170 | |||
AY789064.2 | - | 3 | 116 | |||
BC096694.1 | - | 3 | 143 | 40026979 | ||
BC101977.1 | - | 3 | 143 | 40004312 | ||
BC103693.1 | - | 3 | 143 | 40004314 | ||
BC108918.1 | - | 3 | 134 | 40017300 | ||
BC125147.1 | - | 2 | 116 | |||
BC127084.1 | - | 3 | 149 | 40120468 | ||
BC144566.1 | - | 3 | 221 | 9053094 | ||
BC171788.1 | - | 3 | 221 | 9054442 | ||
L76671.1 | - | 3 | 143 | |||
U24077.1 | - | 3 | 123 | |||
U96188.1 | - | 1 | 69 | |||
X94609.1 | - | 3 | 187 |
Gene Symbol: KIR2DS5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014513.2 | NP_055328.2 | 3 | 166 | ||
GenBank mRNA | AF208054.1 | - | 3 | 166 | ||
AF272389.1 | - | 3 | 116 | |||
AY366247.1 | - | 3 | 190 | |||
AY366248.1 | - | 3 | 190 | |||
L76672.1 | - | 3 | 143 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
