siRNA Details
Gene Symbol: UGT1A7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019077.2 | NP_061950.2 | 1 | 228 | ||
GenBank mRNA | AY435142.1 | - | 1 | 228 | ||
U89507.1 | - | 1 | 228 |
Gene Symbol: UGT1A10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019075.2 | NP_061948.1 | 1 | 274 | ||
GenBank mRNA | AY435137.1 | - | 1 | 228 | ||
BC020971.1 | - | 1 | 297 | 3850654 | ||
BC053576.1 | - | 1 | 249 | 4452440 | ||
BC069210.1 | - | 1 | 258 | 4660540 | ||
U89508.1 | - | 1 | 228 |
Gene Symbol: UGT1A8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019076.4 | NP_061949.3 | 1 | 291 | ||
GenBank mRNA | AF030310.1 | - | 1 | 228 | ||
AF462267.1 | - | 1 | 261 | |||
AF462268.1 | - | 1 | 291 | |||
AK313488.1 | - | 1 | 274 | |||
AY435143.1 | - | 1 | 228 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
