siRNA Details
Gene Symbol: LOC102724197 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006724921.3 | XP_006724984.1 | 9, 10 | 1905 | ||
XM_006724924.3 | XP_006724987.1 | 8, 9 | 1747 | |||
XM_006724928.3 | XP_006724991.1 | 4, 5 | 943 | |||
XM_006724929.2 | XP_006724992.1 | 2, 3 | 292 | |||
XM_011546256.2 | XP_011544558.1 | 10, 11 | 2039 | |||
XM_017030133.1 | XP_016885622.1 | 10, 11 | 1470 | |||
XM_017030134.1 | XP_016885623.1 | 10, 11 | 1473 | |||
XM_017030135.1 | XP_016885624.1 | 10, 11 | 1898 | |||
XM_017030136.1 | XP_016885625.1 | 11, 12 | 1940 | |||
XM_017030137.1 | XP_016885626.1 | 11, 12 | 1937 | |||
XM_017030138.1 | XP_016885627.1 | 11, 12 | 1460 | |||
XM_017030139.1 | XP_016885628.1 | 11, 12 | 1463 | |||
XM_017030140.1 | XP_016885629.1 | 12, 13 | 1930 | |||
XM_017030141.1 | XP_016885630.1 | 12, 13 | 1927 | |||
XM_017030142.1 | XP_016885631.1 | 10, 11 | 1771 | |||
XM_017030143.1 | XP_016885632.1 | 9, 10 | 1906 | |||
XM_017030144.1 | XP_016885633.1 | 5, 6 | 834 | |||
XM_017030145.1 | XP_016885634.1 | 6, 7 | 1467 |
Gene Symbol: GGT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288833.1 | NP_001275762.1 | 12, 13 | 2000 | ||
NM_013421.2 | NP_038265.2 | 13, 14 | 1928 | |||
NM_013430.2 | NP_038347.2 | 12, 13 | 1688 | |||
GenBank mRNA | AL832738.1 | - | 1904 | |||
BC025927.1 | - | 10, 11 | 1568 | 4548861 | ||
BC065921.1 | - | 2, 3 | 211 | |||
BC069326.1 | - | 1, 2 | 169 | |||
BC069473.1 | - | 1, 2 | 169 | |||
BC069504.1 | - | 1, 2 | 169 | |||
BC128234.1 | - | 1, 2 | 197 | |||
BC128238.1 | - | 1, 2 | 198 | |||
BC128239.1 | - | 1, 2 | 198 | |||
CR456494.1 | - | 9, 10 | 1278 | |||
DQ000492.2 | - | 3, 4 | 383 | |||
J04131.1 | - | 13, 14 | 1870 | |||
J05235.1 | - | 11, 12 | 1967 | |||
L20490.1 | - | 2, 3 | 208 | |||
L20493.1 | - | 1, 2 | 357 | |||
M24087.1 | - | 8, 9 | 1203 | |||
M24903.1 | - | 12, 13 | 1688 | |||
X60069.1 | - | 10, 11 | 1559 |
Gene Symbol: GGT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005261890.4 | XP_005261947.1 | 5, 6 | 754 | ||
XM_006724392.3 | XP_006724455.1 | 9, 10 | 1462 | |||
XM_006724395.3 | XP_006724458.1 | 6, 7 | 914 | |||
XM_006724396.2 | XP_006724459.1 | 2, 3 | 292 | |||
XM_011530621.2 | XP_011528923.1 | 2, 3 | 279 | |||
XM_017029179.1 | XP_016884668.1 | 10, 11 | 2143 | |||
XM_017029180.1 | XP_016884669.1 | 9, 10 | 1601 | |||
XM_017029181.1 | XP_016884670.1 | 9, 10 | 2039 | |||
XM_017029182.1 | XP_016884671.1 | 10, 11 | 2143 | |||
XM_017029183.1 | XP_016884672.1 | 5, 6 | 730 | |||
XM_017029184.1 | XP_016884673.1 | 2, 3 | 302 | |||
XM_017029185.1 | XP_016884674.1 | 11, 12 | 3005 | |||
XM_017029186.1 | XP_016884675.1 | 14, 15 | 2606 |
Gene Symbol: GGT3P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003267.1 | 9, 10 | 1395 |
Gene Symbol: GGTLC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178311.2 | NP_842563.1 | 2, 3 | 374 | ||
NM_178312.2 | NP_842564.1 | 2, 3 | 302 | |||
XM_005260863.4 | XP_005260920.1 | 2, 3 | 729 | |||
XM_005260864.4 | XP_005260921.1 | 3, 4 | 882 | |||
XM_005260865.4 | XP_005260922.1 | 3, 4 | 882 | |||
XM_011529391.2 | XP_011527693.1 | 2, 3 | 296 | |||
XM_011529392.2 | XP_011527694.1 | 3, 4 | 821 | |||
XM_011529393.2 | XP_011527695.1 | 2, 3 | 668 | |||
XM_017028126.1 | XP_016883615.1 | 2, 3 | 291 | |||
GenBank mRNA | BC040904.1 | - | 2, 3 | 374 | 5744021 | |
L20491.1 | - | 2, 3 | 302 | |||
L20492.1 | - | 1, 2 | 352 |
Gene Symbol: GGTLC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282879.1 | NP_001269808.1 | 2, 3 | 231 | ||
NM_199127.2 | NP_954578.2 | 1, 2 | 203 | |||
XM_005261815.3 | XP_005261872.2 | 2, 3 | 260 | |||
XM_005261817.2 | XP_005261874.1 | 2, 3 | 235 | |||
XM_005261818.2 | XP_005261875.1 | 2, 3 | 260 | |||
XM_011530497.1 | XP_011528799.1 | 2, 3 | 343 | |||
GenBank mRNA | BC069534.1 | - | 1, 2 | 169 | 7262125 | |
BX282400.1 | - | 2, 3 | 233 | |||
EL736203.1 | - | 1, 2 | 169 | |||
HY050283.1 | - | 2, 3 | 222 | |||
X98922.1 | - | 1, 2 | 204 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
