siRNA Details
Gene Symbol: GGTLC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178311.2 | NP_842563.1 | 3 | 419 | ||
NM_178312.2 | NP_842564.1 | 3 | 347 | |||
XM_005260863.4 | XP_005260920.1 | 3 | 774 | |||
XM_005260864.4 | XP_005260921.1 | 4 | 927 | |||
XM_005260865.4 | XP_005260922.1 | 4 | 927 | |||
XM_011529391.2 | XP_011527693.1 | 3 | 341 | |||
XM_011529392.2 | XP_011527694.1 | 4 | 866 | |||
XM_011529393.2 | XP_011527695.1 | 3 | 713 | |||
XM_017028126.1 | XP_016883615.1 | 3 | 336 | |||
GenBank mRNA | BC040904.1 | - | 3 | 419 | 5744021 | |
L20491.1 | - | 3 | 347 | |||
L20492.1 | - | 2 | 397 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
