siRNA Details
Gene Symbol: PI4KAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003700.1 | 4 | 472 | |||
GenBank mRNA | AK126860.1 | - | 2 | 2436 | ||
AK298744.1 | - | 4 | 493 | |||
AK299734.1 | - | 4 | 337 | |||
BC020225.1 | - | 4 | 470 | |||
BC139841.1 | - | 2 | 193 | |||
CR456378.1 | - | 2 | 211 | |||
DA463001.1 | - | 4 | 416 |
Gene Symbol: PI4KA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_058004.3 | NP_477352.3 | 34 | 4033 | ||
XM_005261634.1 | XP_005261691.1 | 33 | 3940 | |||
XM_005261635.1 | XP_005261692.1 | 34 | 4033 | |||
XM_011530226.1 | XP_011528528.1 | 34 | 4033 | |||
XM_017028829.1 | XP_016884318.1 | 33 | 3967 | |||
XM_017028830.1 | XP_016884319.1 | 20 | 2189 | |||
GenBank mRNA | AB210002.1 | - | 34 | 4003 | ||
AF012872.1 | - | 33 | 3772 | |||
AK302424.1 | - | 34 | 3961 | |||
AK314781.1 | - | 6 | 687 | |||
BC018120.1 | - | 4 | 350 | |||
BC053654.1 | - | 3 | 326 | |||
L36151.1 | - | 2 | 319 |
Gene Symbol: PI4KAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003563.1 | 2 | 205 | |||
GenBank mRNA | AK096724.1 | - | 4 | 431 | ||
AK292695.1 | - | 3 | 296 | |||
BC072398.1 | - | 4 | 481 | |||
BC131538.1 | - | 2 | 205 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
