siRNA Details
Gene Symbol: FCAR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002000.3 | NP_001991.1 | 5 | 957 | ||
NM_133269.3 | NP_579803.1 | 5 | 891 | |||
NM_133271.3 | NP_579805.1 | 4 | 669 | |||
NM_133272.3 | NP_579806.1 | 4 | 921 | |||
NM_133273.3 | NP_579807.1 | 3 | 633 | |||
NM_133274.3 | NP_579808.1 | 4 | 847 | |||
NM_133277.3 | NP_579811.1 | 3 | 630 | |||
NM_133278.3 | NP_579812.1 | 4 | 855 | |||
XM_011526625.2 | XP_011524927.1 | 5 | 881 | |||
XM_017026473.1 | XP_016881962.1 | 5 | 930 | |||
XM_017026474.1 | XP_016881963.1 | 4 | 713 | |||
GenBank mRNA | AK290922.1 | - | 5 | 815 | ||
BC027953.1 | - | 5 | 784 | 5211453 | ||
D87853.1 | - | 5 | 694 | |||
D87854.1 | - | 4 | 472 | |||
D87855.1 | - | 4 | 724 | |||
D87856.1 | - | 3 | 436 | |||
D87857.1 | - | 4 | 650 | |||
D87858.1 | - | 5 | 550 | |||
D87859.1 | - | 6 | 772 | |||
D87860.1 | - | 4 | 514 | |||
D87861.1 | - | 3 | 433 | |||
D87862.1 | - | 4 | 658 | |||
DQ075334.1 | - | 5 | 760 | |||
DQ075335.1 | - | 5 | 760 | |||
DQ075336.1 | - | 5 | 760 | |||
DQ075337.1 | - | 5 | 760 | |||
DQ075338.1 | - | 5 | 760 | |||
DQ075339.1 | - | 5 | 760 | |||
S82919.1 | - | 4 | 472 | |||
U43677.1 | - | 4 | 477 | |||
U43774.1 | - | 5 | 699 | |||
X90753.1 | - | 2 | 221 | |||
X92558.1 | - | 4 | 724 | |||
X92559.1 | - | 4 | 658 | |||
X92560.1 | - | 5 | 694 | |||
X92561.1 | - | 4 | 472 | |||
X92562.1 | - | 3 | 433 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
