siRNA Details
Gene Symbol: PTPRC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002838.4 | NP_002829.3 | 12 | 1443 | ||
NM_080921.3 | NP_563578.2 | 9 | 960 | |||
XM_006711472.3 | XP_006711535.1 | 11 | 1260 | |||
XM_006711473.3 | XP_006711536.1 | 11 | 1188 | |||
XM_006711474.3 | XP_006711537.1 | 10 | 1044 | |||
GenBank mRNA | AK292131.1 | - | 9 | 833 | ||
AK299986.1 | - | 11 | 1118 | |||
AK303539.1 | - | 12 | 1316 | |||
AK307635.1 | - | 11 | 1177 | |||
BC014239.2 | - | 11 | 1163 | |||
Y00062.1 | - | 10 | 887 | |||
Y00638.1 | - | 12 | 1310 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
