siRNA Details
Gene Symbol: BAX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291428.1 | NP_001278357.1 | 5 | 461 | ||
NM_001291429.1 | NP_001278358.1 | 4 | 409 | |||
NM_001291430.1 | NP_001278359.1 | 3 | 262 | |||
NM_001291431.1 | NP_001278360.1 | 3 | 262 | |||
NM_004324.3 | NP_004315.1 | 5 | 461 | |||
NM_138761.3 | NP_620116.1 | 5 | 461 | |||
NM_138763.3 | NP_620118.1 | 4 | 314 | |||
NM_138764.4 | NP_620119.2 | 5 | 461 | |||
NR_027882.1 | 6 | 559 | ||||
XM_017027077.1 | XP_016882566.1 | 5 | 470 | |||
GenBank mRNA | AF007826.1 | - | 6 | 490 | ||
AF247393.1 | - | 5 | 392 | |||
AF250190.1 | - | 3 | 193 | |||
AJ417988.1 | - | 4 | 371 | |||
AJ586909.1 | - | 3 | 180 | |||
AJ586910.1 | - | 4 | 327 | |||
AK291076.1 | - | 5 | 441 | |||
BC014175.2 | - | 5 | 444 | 4578562 | ||
BM706954.1 | - | 3 | 274 | |||
CN281003.1 | - | 5 | 414 | |||
HQ889830.1 | - | 3 | 193 | |||
JX524562.1 | - | 4 | 341 | |||
KF830846.1 | - | 4 | 341 | |||
KF830847.1 | - | 4 | 341 | |||
KF830848.1 | - | 4 | 341 | |||
L22473.1 | - | 5 | 392 | |||
L22474.1 | - | 5 | 392 | |||
T29044.1 | - | 2 | 129 | |||
U19599.1 | - | 4 | 245 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
