siRNA Details
Gene Symbol: BAX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291429.1 | NP_001278358.1 | 4 | 740 | ||
NM_001291430.1 | NP_001278359.1 | 3 | 593 | |||
NM_004324.3 | NP_004315.1 | 5 | 792 | |||
GenBank mRNA | BM706954.1 | - | 3 | 606 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
