siRNA Details
Gene Symbol: INPP5F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243195.1 | NP_001230124.1 | 1, 2 | 318 | ||
NM_014937.3 | NP_055752.1 | 1, 2 | 318 | |||
XM_006717720.3 | XP_006717783.1 | 1, 2 | 2705 | |||
XM_011539525.2 | XP_011537827.1 | 1, 2 | 2705 | |||
XM_017015936.1 | XP_016871425.1 | 1, 2 | 226 | |||
GenBank mRNA | AB023183.1 | - | 1, 2 | 268 | ||
AK307758.1 | - | 1, 2 | 308 | |||
BC052367.1 | - | 1, 2 | 203 | |||
BC067820.1 | - | 1, 2 | 279 | |||
BC082755.1 | - | 1, 2 | 203 | |||
BC111493.1 | - | 1, 2 | 229 | 6166990 | ||
DB636424.1 | - | 1, 2 | 317 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
