siRNA Details
Gene Symbol: INPP5F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243195.1 | NP_001230124.1 | 4 | 622 | ||
NM_014937.3 | NP_055752.1 | 4 | 622 | |||
XM_006717720.3 | XP_006717783.1 | 4 | 3009 | |||
XM_011539524.2 | XP_011537826.1 | 4 | 385 | |||
XM_011539525.2 | XP_011537827.1 | 5 | 3092 | |||
XM_011539527.2 | XP_011537829.2 | 3 | 387 | |||
XM_011539528.2 | XP_011537830.1 | 3 | 509 | |||
XM_017015935.1 | XP_016871424.1 | 4 | 376 | |||
XM_017015936.1 | XP_016871425.1 | 7 | 1010 | |||
GenBank mRNA | AB023183.1 | - | 4 | 572 | ||
AK307758.1 | - | 4 | 612 | |||
BC052367.1 | - | 4 | 507 | |||
BC067820.1 | - | 4 | 583 | |||
BC082755.1 | - | 4 | 507 | |||
BC111493.1 | - | 4 | 533 | 6166990 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
