siRNA Details
Gene Symbol: WWOX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016373.3 | NP_057457.1 | 2, 3 | 543 | ||
NM_130791.3 | NP_570607.1 | 2, 3 | 543 | |||
NR_120436.1 | 2, 3 | 657 | ||||
XM_011523101.2 | XP_011521403.1 | 2, 3 | 302 | |||
XM_011523103.2 | XP_011521405.1 | 2, 3 | 302 | |||
XM_011523104.2 | XP_011521406.1 | 2, 3 | 302 | |||
XM_011523105.2 | XP_011521407.1 | 2, 3 | 302 | |||
XM_017023278.1 | XP_016878767.1 | 2, 3 | 302 | |||
GenBank mRNA | AF187015.1 | - | 2, 3 | 261 | ||
AF211943.1 | - | 2, 3 | 302 | |||
AF227526.1 | - | 2, 3 | 262 | |||
AF227527.1 | - | 2, 3 | 281 | |||
AF227528.1 | - | 2, 3 | 278 | |||
AF395123.1 | - | 2, 3 | 310 | |||
AF395124.1 | - | 2, 3 | 310 | |||
AK027626.1 | - | 2, 3 | 301 | |||
AK290438.1 | - | 2, 3 | 293 | |||
AK291300.1 | - | 2, 3 | 300 | |||
AW874692.1 | - | 2, 3 | 188 | |||
AY256821.1 | - | 2, 3 | 232 | |||
BC003184.1 | - | 2, 3 | 256 | 3535809 | ||
BT007445.1 | - | 177 | ||||
BU157392.1 | - | 2, 3 | 284 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
