siRNA Details
Gene Symbol: DNMT3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320893.1 | NP_001307822.1 | 17 | 2084 | ||
NM_022552.4 | NP_072046.2 | 22 | 2770 | |||
NM_153759.3 | NP_715640.2 | 18 | 2054 | |||
NM_175629.2 | NP_783328.1 | 22 | 2841 | |||
NR_135490.1 | 23 | 3040 | ||||
XM_005264175.4 | XP_005264232.1 | 22 | 2704 | |||
XM_005264177.4 | XP_005264234.1 | 17 | 1996 | |||
XM_011532662.2 | XP_011530964.1 | 20 | 2706 | |||
XM_011532663.2 | XP_011530965.1 | 23 | 2883 | |||
XM_011532666.2 | XP_011530968.1 | 18 | 2069 | |||
XM_011532667.2 | XP_011530969.1 | 17 | 1886 | |||
XM_017003526.1 | XP_016859015.1 | 22 | 2812 | |||
XM_017003527.1 | XP_016859016.1 | 17 | 2040 | |||
GenBank mRNA | AB208833.1 | - | 23 | 2939 | ||
AF067972.2 | - | 22 | 2732 | |||
AF331856.1 | - | 22 | 2720 | |||
AF480163.1 | - | 18 | 2050 | |||
AK025230.1 | - | 12 | 1094 | |||
BC018214.1 | - | 7 | 623 | |||
BC023612.2 | - | 8 | 822 | |||
BC043617.1 | - | 22 | 2740 | 6150112 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
