siRNA Details
Gene Symbol: BCL2L11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204106.1 | NP_001191035.1 | 4 | 2240 | ||
NM_001204107.1 | NP_001191036.1 | 6 | 2437 | |||
NM_001204108.1 | NP_001191037.1 | 5 | 2642 | |||
NM_001204109.1 | NP_001191038.1 | 4 | 2531 | |||
NM_001204110.1 | NP_001191039.1 | 5 | 2372 | |||
NM_001204111.1 | NP_001191040.1 | 4 | 2226 | |||
NM_001204112.1 | NP_001191041.1 | 5 | 2351 | |||
NM_006538.4 | NP_006529.1 | 5 | 2330 | |||
NM_138621.4 | NP_619527.1 | 4 | 2510 | |||
NM_138622.3 | NP_619528.1 | 5 | 2635 | |||
NM_138623.3 | NP_619529.1 | 6 | 2455 | |||
NM_138624.3 | NP_619530.1 | 5 | 2543 | |||
NM_138625.3 | NP_619531.1 | 5 | 2356 | |||
NM_138626.3 | NP_619532.1 | 3 | 2406 | |||
NM_138627.3 | NP_619533.1 | 3 | 2136 | |||
NM_207003.2 | NP_996886.1 | 5 | 2365 | |||
XM_005263551.2 | XP_005263608.1 | 4 | 3288 | |||
XM_005263553.2 | XP_005263610.1 | 5 | 3108 | |||
XM_005263554.2 | XP_005263611.1 | 3 | 3184 | |||
XM_005263559.2 | XP_005263616.1 | 4 | 3018 | |||
XM_017003101.1 | XP_016858590.1 | 4 | 3004 | |||
GenBank mRNA | AK027160.1 | - | 1 | 945 | ||
BC033694.1 | - | 4 | 2492 | 5213713 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
