siRNA Details
Gene Symbol: ARHGAP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198726.1 | NP_001185655.1 | 4, 5 | 434 | ||
NM_181335.2 | NP_851852.2 | 4, 5 | 434 | |||
GenBank mRNA | AK022305.1 | - | 3, 4 | 376 | ||
BC010490.2 | - | 2, 3 | 278 | |||
BC048280.1 | - | 4, 5 | 386 | 5190702 | ||
BC059382.1 | - | 4, 5 | 418 |
Gene Symbol: PRR5-ARHGAP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181334.5 | NP_851851.3 | 7, 8 | 963 | ||
GenBank mRNA | AF195968.1 | - | 6, 7 | 554 | ||
BC132755.1 | - | 5, 6 | 454 | 40146777 | ||
BC132757.1 | - | 5, 6 | 454 | 40146779 | ||
BC144382.1 | - | 5, 6 | 454 | 9052903 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
