siRNA Details
Gene Symbol: DLC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024767.3 | NP_079043.3 | 2 | 540 | ||
NM_182643.2 | NP_872584.2 | 2 | 540 | |||
XM_005273374.1 | XP_005273431.1 | 2 | 421 | |||
XM_017012951.1 | XP_016868440.1 | 2 | 291 | |||
GenBank mRNA | AB051510.1 | - | 2 | 426 | ||
AK024773.1 | - | 2 | 385 | |||
AK225098.1 | - | 2 | 385 | |||
AK291682.1 | - | 2 | 381 | |||
BC049842.1 | - | 2 | 507 | |||
BC054511.1 | - | 2 | 295 | 5240161 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
