siRNA Details
Gene Symbol: TBC1D25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002536.2 | NP_002527.1 | 3, 4 | 503 | ||
GenBank mRNA | AB449912.1 | - | 3, 4 | 392 | ||
AK293875.1 | - | 3, 4 | 462 | |||
AK294776.1 | - | 3, 4 | 401 | |||
BC101817.1 | - | 3, 4 | 429 | 8069323 | ||
BC101819.1 | - | 3, 4 | 429 | 8069325 | ||
BC125088.1 | - | 3, 4 | 418 | 40117866 | ||
L08240.1 | - | 2, 3 | 282 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
