siRNA Details
Gene Symbol: OR14J1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030946.1 | NP_112208.1 | 1 | 815 | ||
GenBank mRNA | BC136905.1 | - | 1 | 879 | 9020896 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
