siRNA Details
Gene Symbol: KLK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001030047.1 | NP_001025218.1 | 5 | 1592 | ||
NM_001030048.1 | NP_001025219.1 | 5 | 1021 | |||
NM_001648.2 | NP_001639.1 | 5 | 1150 | |||
GenBank mRNA | AJ310938.1 | - | 4 | 1289 | ||
BC005307.1 | - | 5 | 1150 | 3950475 | ||
BC050595.2 | - | 5 | 1173 | 6199928 | ||
BC056665.1 | - | 5 | 1155 | 4296876 | ||
M21895.1 | - | 5 | 1099 | |||
M21896.1 | - | 5 | 1541 | |||
M26663.1 | - | 5 | 1133 | |||
X05332.1 | - | 5 | 1152 | |||
X07730.1 | - | 4 | 1414 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
