siRNA Details
Gene Symbol: BAX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291428.1 | NP_001278357.1 | 3 | 281 | ||
NM_001291429.1 | NP_001278358.1 | 2 | 229 | |||
NM_004324.3 | NP_004315.1 | 3 | 281 | |||
NM_138761.3 | NP_620116.1 | 3 | 281 | |||
NM_138764.4 | NP_620119.2 | 3 | 281 | |||
NR_027882.1 | 3 | 281 | ||||
XM_017027077.1 | XP_016882566.1 | 3 | 290 | |||
GenBank mRNA | AF007826.1 | - | 3 | 212 | ||
AF247393.1 | - | 3 | 212 | |||
AJ417988.1 | - | 2 | 191 | |||
AJ586910.1 | - | 2 | 147 | |||
AK291076.1 | - | 3 | 261 | |||
BC014175.2 | - | 3 | 264 | 4578562 | ||
BE396495.1 | - | 3 | 281 | |||
BM706954.1 | - | 1 | 94 | |||
CN281003.1 | - | 3 | 234 | |||
JX524562.1 | - | 2 | 161 | |||
KF830846.1 | - | 2 | 161 | |||
KF830847.1 | - | 2 | 161 | |||
KF830848.1 | - | 2 | 161 | |||
L22473.1 | - | 3 | 212 | |||
L22474.1 | - | 3 | 212 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
