siRNA Details
Gene Symbol: KIR3DL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153443.4 | NP_703144.3 | 3, 4 | 416 | ||
GenBank mRNA | AF352324.1 | - | 3, 4 | 384 | ||
AJ938052.1 | - | 3, 4 | 371 | |||
AJ938053.1 | - | 3, 4 | 371 | |||
AJ938054.1 | - | 3, 4 | 371 | |||
AJ938055.1 | - | 3, 4 | 371 | |||
AJ938056.1 | - | 3, 4 | 371 | |||
AJ938057.1 | - | 3, 4 | 371 | |||
AJ938058.1 | - | 3, 4 | 371 | |||
AJ938059.1 | - | 3, 4 | 371 | |||
AJ938060.1 | - | 3, 4 | 371 | |||
AJ938061.1 | - | 3, 4 | 371 | |||
AJ938062.1 | - | 3, 4 | 371 | |||
AJ938063.1 | - | 3, 4 | 371 | |||
AJ938064.1 | - | 3, 4 | 371 | |||
AJ938065.1 | - | 3, 4 | 371 | |||
AJ938066.1 | - | 3, 4 | 371 | |||
BC136283.1 | - | 3, 4 | 416 | 9020270 | ||
BC143802.1 | - | 3, 4 | 416 | 9052316 |
Gene Symbol: 1060P11.3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AJ630586.1 | - | 3, 4 | 367 |
Gene Symbol: KIR2DL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014218.2 | NP_055033.2 | 2, 3 | 119 | ||
XM_011526939.2 | XP_011525241.1 | 2, 3 | 724 | |||
XM_017026782.1 | XP_016882271.1 | 2, 3 | 97 | |||
XM_017026783.1 | XP_016882272.1 | 2, 3 | 724 | |||
GenBank mRNA | AF022045.1 | - | 2, 3 | 88 | ||
AF285431.1 | - | 2, 3 | 114 | |||
AF285432.1 | - | 1, 2 | 41 | |||
AY366237.1 | - | 2, 3 | 117 | |||
AY366238.1 | - | 2, 3 | 63 | |||
AY366239.1 | - | 2, 3 | 111 | |||
AY366240.1 | - | 2, 3 | 119 | |||
AY366241.1 | - | 2, 3 | 118 | |||
AY388471.1 | - | 2, 3 | 111 | |||
AY789055.1 | - | 2, 3 | 114 | |||
AY789056.1 | - | 2, 3 | 81 | |||
BC069344.1 | - | 2, 3 | 114 | 7262296 | ||
KJ699235.1 | - | 2, 3 | 354 | |||
L41267.1 | - | 2, 3 | 113 | |||
U24076.1 | - | 2, 3 | 114 | |||
U24078.1 | - | 2, 3 | 121 |
Gene Symbol: KIR2DL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014219.2 | NP_055034.2 | 2, 3 | 110 | ||
GenBank mRNA | AF285434.1 | - | 1, 2 | 41 | ||
AY366242.1 | - | 2, 3 | 110 | |||
KP260924.1 | - | 2, 3 | 77 | |||
L76669.1 | - | 2, 3 | 92 | |||
U24075.1 | - | 2, 3 | 95 |
Gene Symbol: KIR2DL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015868.2 | NP_056952.2 | 2, 3 | 118 | ||
GenBank mRNA | AF022048.1 | - | 2, 3 | 81 | ||
AF135556.1 | - | 1 | 18 | |||
AF135566.1 | - | 1 | 18 | |||
AF135567.1 | - | 1 | 18 | |||
AF285435.1 | - | 1, 2 | 41 | |||
AY789057.1 | - | 2, 3 | 81 | |||
BC032422.1 | - | 2, 3 | 94 | 5220339 | ||
BC050730.1 | - | 2, 3 | 112 | 5219336 | ||
BC110613.1 | - | 2, 3 | 95 | 40031619 | ||
FN395076.1 | - | 2, 3 | 81 | |||
FN552550.1 | - | 2, 3 | 63 | |||
L41268.1 | - | 2, 3 | 118 | |||
L76662.1 | - | 2, 3 | 92 | |||
L76663.1 | - | 2, 3 | 92 | |||
U24074.1 | - | 2, 3 | 102 | |||
U50546.1 | - | 2, 3 | 92 | |||
U73395.1 | - | 2, 3 | 102 |
Gene Symbol: KIR2DS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014512.1 | NP_055327.1 | 2, 3 | 94 | ||
GenBank mRNA | AF022046.1 | - | 2, 3 | 94 | ||
AF285437.1 | - | 1, 2 | 41 | |||
AM999888.1 | - | 2, 3 | 89 | |||
X89892.1 | - | 2, 3 | 101 | |||
X98858.1 | - | 2, 3 | 81 |
Gene Symbol: KIR2DS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291695.1 | NP_001278624.1 | 2, 3 | 116 | ||
NM_001291701.1 | NP_001278630.1 | 2, 3 | 116 | |||
NM_012312.4 | NP_036444.1 | 2, 3 | 116 | |||
GenBank mRNA | AF135565.1 | - | 1 | 18 | ||
AF285438.1 | - | 1, 2 | 41 | |||
AJ002102.1 | - | 2, 3 | 81 | |||
AJ002103.1 | - | 2, 3 | 81 | |||
AJ002104.1 | - | 2, 3 | 81 | |||
AJ002105.1 | - | 2, 3 | 81 | |||
AM409094.1 | - | 2, 3 | 82 | |||
AY366243.1 | - | 2, 3 | 91 | |||
AY366244.1 | - | 2, 3 | 80 | |||
BC108917.1 | - | 2, 3 | 83 | 40017299 | ||
BC119631.1 | - | 2, 3 | 100 | 40115568 | ||
BC119632.1 | - | 2, 3 | 100 | 40115570 | ||
BC125146.1 | - | 1, 2 | 65 | |||
BC126856.1 | - | 2, 3 | 83 | 40017301 | ||
FR750994.1 | - | 2, 3 | 116 | |||
L41347.1 | - | 2, 3 | 100 | |||
L76667.1 | - | 2, 3 | 92 | |||
U24079.1 | - | 2, 3 | 79 | |||
X89893.1 | - | 2, 3 | 81 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
