siRNA Details
Gene Symbol: KIR3DL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322168.1 | NP_001309097.1 | 5 | 884 | ||
NM_013289.2 | NP_037421.2 | 5 | 884 | |||
GenBank mRNA | AF022049.1 | - | 5 | 821 | ||
AF103010.1 | - | 5 | 821 | |||
AF103011.1 | - | 5 | 821 | |||
AF103012.1 | - | 5 | 821 | |||
AF103013.1 | - | 5 | 821 | |||
AF208688.1 | - | 3 | 758 | |||
AF208689.1 | - | 3 | 758 | |||
AF262968.1 | - | 3 | 740 | |||
AF262969.1 | - | 3 | 740 | |||
AF262970.1 | - | 3 | 740 | |||
AF262971.1 | - | 3 | 740 | |||
AF262972.1 | - | 3 | 740 | |||
AF262973.1 | - | 5 | 854 | |||
AF262974.1 | - | 5 | 854 | |||
AJ417556.1 | - | 3 | 770 | |||
AJ417557.1 | - | 3 | 770 | |||
AY366236.1 | - | 5 | 859 | |||
AY366249.1 | - | 5 | 821 | |||
AY366250.1 | - | 5 | 847 | |||
AY366251.1 | - | 5 | 821 | |||
AY366252.1 | - | 5 | 821 | |||
AY760050.1 | - | 5 | 821 | |||
AY789065.1 | - | 5 | 836 | |||
AY789066.1 | - | 5 | 821 | |||
AY789072.1 | - | 5 | 821 | |||
BC028206.1 | - | 5 | 848 | 5215674 | ||
BC101540.1 | - | 5 | 840 | 8069046 | ||
BC101542.1 | - | 5 | 840 | 8069048 | ||
DQ178659.1 | - | 5 | 821 | |||
EF462885.1 | - | 3 | 751 | |||
EF472674.1 | - | 3 | 751 | |||
EF472675.1 | - | 3 | 751 | |||
EF472676.1 | - | 3 | 751 | |||
EF472677.1 | - | 3 | 751 | |||
EF472679.1 | - | 3 | 751 | |||
L41269.1 | - | 5 | 840 | |||
L76664.1 | - | 4 | 547 | |||
U30273.1 | - | 5 | 823 | |||
U30274.1 | - | 5 | 884 | |||
U31416.1 | - | 5 | 846 | |||
U33328.1 | - | 5 | 846 | |||
X94262.1 | - | 5 | 822 |
Gene Symbol: KIR2DS5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014513.2 | NP_055328.2 | 4 | 570 | ||
GenBank mRNA | AF208054.1 | - | 4 | 570 | ||
L76672.1 | - | 4 | 547 |
Gene Symbol: KIR3DL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242867.1 | NP_001229796.1 | 5 | 854 | ||
NM_006737.3 | NP_006728.2 | 5 | 854 | |||
XM_011526940.2 | XP_011525242.1 | 5 | 978 | |||
XM_017026784.1 | XP_016882273.1 | 5 | 978 | |||
GenBank mRNA | AF208683.1 | - | 3 | 758 | ||
AF208684.1 | - | 3 | 758 | |||
AF208685.1 | - | 3 | 758 | |||
AF208686.1 | - | 3 | 758 | |||
AF262965.1 | - | 3 | 740 | |||
AF262966.1 | - | 3 | 740 | |||
AF262967.1 | - | 5 | 854 | |||
AF263617.1 | - | 5 | 854 | |||
AJ276125.1 | - | 5 | 822 | |||
AW951147.1 | - | 2 | 479 | |||
AY059418.1 | - | 3 | 752 | |||
AY059419.1 | - | 3 | 752 | |||
AY059420.1 | - | 3 | 752 | |||
AY366256.2 | - | 5 | 833 | |||
AY789067.1 | - | 5 | 821 | |||
AY789068.1 | - | 5 | 821 | |||
AY789069.1 | - | 4 | 791 | |||
AY789070.1 | - | 5 | 821 | |||
AY789071.1 | - | 4 | 791 | |||
BC105678.1 | - | 5 | 846 | 40004817 | ||
BC107951.1 | - | 5 | 846 | 40004820 | ||
DQ256504.1 | - | 3 | 753 | |||
EF582384.1 | - | 5 | 821 | |||
L41270.1 | - | 5 | 840 | |||
L76665.1 | - | 5 | 832 | |||
L76666.1 | - | 5 | 832 | |||
U30272.1 | - | 5 | 823 | |||
X93595.1 | - | 5 | 851 | |||
X93596.1 | - | 5 | 821 | |||
X94373.1 | - | 5 | 822 | |||
X94374.1 | - | 5 | 821 |
Gene Symbol: KIR3DS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083539.2 | NP_001077008.1 | 5 | 858 | ||
NM_001282170.1 | NP_001269099.1 | 4 | 573 | |||
NM_001282171.1 | NP_001269100.1 | 3 | 537 | |||
GenBank mRNA | AF022044.1 | - | 5 | 834 | ||
AJ417558.3 | - | 3 | 770 | |||
AK301140.1 | - | 5 | 858 | |||
BC098107.1 | - | 3 | 511 | 40026977 | ||
BC098288.1 | - | 5 | 832 | 40026980 | ||
BC105679.1 | - | 5 | 832 | 40004313 | ||
EF370406.1 | - | 4 | 781 | |||
EF582383.1 | - | 5 | 821 | |||
L76661.1 | - | 5 | 832 | |||
U73396.1 | - | 5 | 821 | |||
X97232.1 | - | 4 | 536 | |||
X97233.1 | - | 5 | 821 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
