siRNA Details
Gene Symbol: KIR2DS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291695.1 | NP_001278624.1 | 3 | 211 | ||
NM_001291701.1 | NP_001278630.1 | 3 | 211 | |||
NM_012312.4 | NP_036444.1 | 3 | 211 | |||
GenBank mRNA | AF285438.1 | - | 2 | 136 | ||
AJ002102.1 | - | 3 | 176 | |||
AJ002103.1 | - | 3 | 176 | |||
AJ002104.1 | - | 3 | 176 | |||
AJ002105.1 | - | 3 | 176 | |||
AM409094.1 | - | 3 | 177 | |||
AY366243.1 | - | 3 | 186 | |||
AY366244.1 | - | 3 | 175 | |||
BC108917.1 | - | 3 | 178 | 40017299 | ||
BC119631.1 | - | 3 | 195 | 40115568 | ||
BC119632.1 | - | 3 | 195 | 40115570 | ||
BC125146.1 | - | 2 | 160 | |||
BC126856.1 | - | 3 | 178 | 40017301 | ||
FR750994.1 | - | 3 | 211 | |||
L41347.1 | - | 3 | 195 | |||
L76667.1 | - | 3 | 187 | |||
U24079.1 | - | 3 | 174 | |||
X89893.1 | - | 3 | 176 |
Gene Symbol: 1060P11.3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AJ630586.1 | - | 4 | 462 |
Gene Symbol: KIR2DL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014219.2 | NP_055034.2 | 3 | 205 | ||
GenBank mRNA | AF285434.1 | - | 2 | 136 | ||
AY366242.1 | - | 3 | 205 | |||
KP260924.1 | - | 3 | 172 | |||
L76669.1 | - | 3 | 187 | |||
U24075.1 | - | 3 | 190 |
Gene Symbol: KIR2DL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF022048.1 | - | 3 | 176 | ||
FN552550.1 | - | 3 | 158 | |||
U73395.1 | - | 3 | 197 |
Gene Symbol: KIR2DS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012313.1 | NP_036445.1 | 3 | 187 | ||
GenBank mRNA | AF022047.1 | - | 3 | 180 | ||
AM851002.1 | - | 3 | 212 | |||
AY789063.2 | - | 2 | 160 | |||
L76670.1 | - | 3 | 187 | |||
X97231.1 | - | 3 | 235 |
Gene Symbol: KIR2DS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281971.1 | NP_001268900.1 | 3 | 265 | ||
NM_001281972.1 | NP_001268901.1 | 3 | 265 | |||
NM_012314.5 | NP_036446.3 | 3 | 265 | |||
GenBank mRNA | AF002255.1 | - | 3 | 222 | ||
AF135563.1 | - | 1 | 113 | |||
AF135564.1 | - | 1 | 113 | |||
AJ417554.1 | - | 2 | 156 | |||
AJ417555.1 | - | 2 | 156 | |||
AJ620615.2 | - | 2 | 149 | |||
AJ620616.2 | - | 2 | 149 | |||
AY102623.1 | - | 3 | 176 | |||
AY366245.1 | - | 3 | 210 | |||
AY366246.1 | - | 3 | 183 | |||
AY388472.1 | - | 3 | 214 | |||
AY789064.2 | - | 3 | 160 | |||
BC096694.1 | - | 3 | 187 | 40026979 | ||
BC101977.1 | - | 3 | 187 | 40004312 | ||
BC103693.1 | - | 3 | 187 | 40004314 | ||
BC108918.1 | - | 3 | 178 | 40017300 | ||
BC125147.1 | - | 2 | 160 | |||
BC127084.1 | - | 3 | 193 | 40120468 | ||
BC144566.1 | - | 3 | 265 | 9053094 | ||
BC171788.1 | - | 3 | 265 | 9054442 | ||
L76671.1 | - | 3 | 187 | |||
U24077.1 | - | 3 | 167 | |||
U96188.1 | - | 1 | 113 | |||
X94609.1 | - | 3 | 231 |
Gene Symbol: KIR2DS5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014513.2 | NP_055328.2 | 3 | 210 | ||
GenBank mRNA | AF208054.1 | - | 3 | 210 | ||
AF272389.1 | - | 3 | 160 | |||
AY366247.1 | - | 3 | 234 | |||
AY366248.1 | - | 3 | 234 | |||
L76672.1 | - | 3 | 187 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
